Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106054.1 | NP_001099524.1 | 7 | 2148 | ||
XM_008770864.1 | XP_008769086.1 | 9 | 2692 |