Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001069.2 | NP_001060.1 | 4 | 1483 | ||
NM_001310315.1 | NP_001297244.1 | 4 | 1615 | |||
GenBank mRNA | AL038778.2 | - | 94 | |||
BC001194.1 | - | 4 | 1446 | 3355410 | ||
BC018780.1 | - | 4 | 1457 | 4870519 | ||
X79535.1 | - | 4 | 1460 |