Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034105.1 | NP_001029277.1 | 5 | 412 | ||
XM_008770987.1 | XP_008769209.1 | 6 | 556 | |||
GenBank mRNA | DQ062205.1 | - | 5 | 412 |