Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Use1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169100.1 | NP_001162571.1 | 1 | 114 | ||
XM_006252855.2 | XP_006252917.1 | 1 | 1034 | |||
GenBank mRNA | BC158812.1 | - | 1 | 90 | ||
FQ217580.1 | - | 1 | 115 |