Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Use1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169100.1 | NP_001162571.1 | 2, 3 | 187 | ||
XM_006252855.2 | XP_006252917.1 | 2, 3 | 1107 | |||
GenBank mRNA | BC158812.1 | - | 2, 3 | 163 | ||
FQ217580.1 | - | 2, 3 | 188 |