Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Babam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006964.1 | NP_001006965.1 | 9 | 1033 | ||
XM_006252857.2 | XP_006252919.1 | 9 | 1057 | |||
GenBank mRNA | BC083685.1 | - | 9 | 1033 | 7189038 | |
FQ214886.1 | - | 9 | 1024 |