Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126337.1 | NP_001119809.1 | 4 | 357 | ||
NM_001301317.1 | NP_001288246.1 | 6 | 567 | |||
NM_020127.2 | NP_064512.1 | 5 | 432 | |||
XM_011509961.2 | XP_011508263.1 | 3 | 266 | |||
XM_017002223.1 | XP_016857712.1 | 5 | 508 | |||
XM_017002224.1 | XP_016857713.1 | 4 | 629 | |||
GenBank mRNA | AF254260.1 | - | 5 | 432 | ||
AK075001.1 | - | 4 | 354 | |||
AK223161.1 | - | 5 | 428 | |||
AK298066.1 | - | 6 | 567 | |||
AK299263.1 | - | 4 | 355 | |||
AK299925.1 | - | 4 | 345 | |||
AK315414.1 | - | 5 | 428 | |||
AL136917.1 | - | 5 | 420 | |||
AY358610.1 | - | 4 | 351 | |||
BC002933.1 | - | 4 | 325 | 3956873 | ||
BC008301.1 | - | 5 | 412 | 3506004 |