Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106070.1 | NP_001099540.1 | 2, 3 | 257 | ||
XM_008771085.1 | XP_008769307.1 | 2, 3 | 240 | |||
XM_008771086.1 | XP_008769308.1 | 2, 3 | 240 | |||
GenBank mRNA | BC169058.1 | - | 2, 3 | 229 | 7932017 | |
FQ211658.1 | - | 2, 3 | 245 |