Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cope | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106076.1 | NP_001099546.1 | 3, 4 | 502 | ||
XM_006252876.2 | XP_006252938.1 | 3, 4 | 516 | |||
GenBank mRNA | BC161928.1 | - | 3, 4 | 516 | 8374450 | |
FQ220757.1 | - | 3, 4 | 326 | |||
FQ232202.1 | - | 3, 4 | 325 | |||
FQ233163.1 | - | 3, 4 | 325 |