Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169119.1 | NP_001162590.1 | 1492 | |||
GenBank mRNA | BC085958.1 | - | 1492 |