Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erlin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106088.2 | NP_001099558.1 | 5 | 320 | ||
XM_008771323.1 | XP_008769545.1 | 5 | 573 | |||
XM_008771324.1 | XP_008769546.1 | 5 | 515 | |||
GenBank mRNA | BC168668.1 | - | 5 | 320 | 7934033 |