Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC290876 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037182.1 | NP_001032259.1 | 1 | 248 | ||
XM_006253430.2 | XP_006253492.1 | 1 | 573 | |||
XM_006253432.2 | XP_006253494.1 | 1 | 574 | |||
XM_008771386.1 | XP_008769608.1 | 1 | 574 | |||
GenBank mRNA | BC085810.1 | - | 1 | 207 | ||
BC100249.1 | - | 1 | 248 | 7135908 |