Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003328.2 | NP_003319.2 | 4 | 386 | ||
XM_011513747.2 | XP_011512049.1 | 4 | 404 | |||
XM_017008580.1 | XP_016864069.1 | 5 | 1588 | |||
XM_017008581.1 | XP_016864070.1 | 3 | 1397 | |||
GenBank mRNA | DB103575.1 | - | 4 | 380 | ||
L27071.1 | - | 4 | 386 |