Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003328.2 | NP_003319.2 | 2, 3 | 155 | ||
GenBank mRNA | DB103575.1 | - | 2, 3 | 149 | ||
L27071.1 | - | 2, 3 | 155 |