Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eci3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009275.1 | NP_001009275.1 | 9 | 931 | ||
GenBank mRNA | BC088178.1 | - | 9 | 931 | 7324966 | |
FQ223513.1 | - | 8 | 771 |