Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYRO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006293.3 | NP_006284.2 | 14 | 1920 | ||
XM_017022542.1 | XP_016878031.1 | 14 | 1885 | |||
XM_017022543.1 | XP_016878032.1 | 14 | 1721 | |||
GenBank mRNA | AB209431.1 | - | 14 | 1949 | ||
AK295496.1 | - | 3 | 215 | |||
BC029925.2 | - | 6 | 570 | |||
BC034960.1 | - | 14 | 1702 | |||
BC049368.1 | - | 14 | 1694 | 4821587 | ||
BC051756.1 | - | 14 | 1699 | 6095596 | ||
D17517.1 | - | 14 | 1908 | |||
D50479.1 | - | 14 | 1680 | |||
U02566.1 | - | 11 | 1131 | |||
U05682.1 | - | 14 | 1690 | |||
X72886.1 | - | 3 | 130 |