Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022851.1 | NP_074042.1 | 2 | 360 | ||
GenBank mRNA | FM032642.1 | - | 2 | 360 | ||
L34821.1 | - | 2 | 239 |