Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000550.2 | NP_000541.1 | 4, 5 | 1084 | ||
GenBank mRNA | AK297887.1 | - | 2, 3 | 326 | ||
BC052608.1 | - | 3, 4 | 1003 | 6258285 | ||
CR407683.1 | - | 3, 4 | 906 | |||
X51420.1 | - | 4, 5 | 1035 | |||
X51455.1 | - | 2, 3 | 257 |