Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106124.1 | NP_001099594.1 | 6 | 1109 | ||
XM_006254285.2 | XP_006254347.1 | 8 | 1745 |