Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106128.1 | NP_001099598.1 | 5 | 531 | ||
XM_006253800.2 | XP_006253862.1 | 6 | 703 |