Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012740.1 | NP_001012758.1 | 2 | 624 | ||
XM_006255022.2 | XP_006255084.1 | 4 | 1125 | |||
XM_006255023.2 | XP_006255085.1 | 3 | 1080 | |||
XM_006255024.2 | XP_006255086.1 | 3 | 760 | |||
GenBank mRNA | BC091341.1 | - | 2 | 624 | 7315458 |