Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282161.1 | NP_001269090.1 | 6 | 636 | ||
NM_003336.3 | NP_003327.2 | 6 | 643 | |||
NM_181762.2 | NP_861427.1 | 5 | 553 | |||
GenBank mRNA | AK223045.1 | - | 6 | 545 | ||
AK223064.1 | - | 6 | 542 | |||
AK297696.1 | - | 5 | 455 | |||
AK301675.1 | - | 5 | 554 | |||
AK313092.1 | - | 6 | 547 | |||
BC010175.2 | - | 6 | 536 | 3051041 | ||
BC042021.1 | - | 3 | 1420 | |||
BI093289.1 | - | 5 | 343 | |||
DA068506.1 | - | 6 | 540 | |||
M74524.1 | - | 6 | 518 |