Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003339.2 | NP_003330.1 | 5 | 919 | ||
NM_181838.1 | NP_862821.1 | 6 | 1083 | |||
XM_017009820.1 | XP_016865309.1 | 8 | 1290 | |||
GenBank mRNA | AF317220.1 | - | 3 | 227 | ||
AK001311.1 | - | 5 | 692 | |||
AK001428.1 | - | 5 | 746 | |||
AK310273.1 | - | 5 | 648 | |||
AY651263.1 | - | 5 | 675 | |||
BC019052.2 | - | 5 | 703 | |||
BC033349.1 | - | 5 | 919 | 4827210 | ||
U39317.1 | - | 5 | 315 |