Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003339.2 | NP_003330.1 | 6 | 1012 | ||
NM_181838.1 | NP_862821.1 | 7 | 1176 | |||
XM_017009820.1 | XP_016865309.1 | 9 | 1383 | |||
GenBank mRNA | AF317220.1 | - | 4 | 320 | ||
AK001311.1 | - | 6 | 785 | |||
AK001428.1 | - | 6 | 839 | |||
AK310273.1 | - | 6 | 741 | |||
AY651263.1 | - | 6 | 768 | |||
BC019052.2 | - | 6 | 796 | |||
BC033349.1 | - | 6 | 1012 | 4827210 | ||
U39317.1 | - | 6 | 408 |