Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oacyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106139.1 | NP_001099609.1 | 10 | 1362 | ||
XM_006254859.2 | XP_006254921.1 | 8 | 1131 |