Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003339.2 | NP_003330.1 | 6 | 999 | ||
NM_181838.1 | NP_862821.1 | 7 | 1163 | |||
XM_017009820.1 | XP_016865309.1 | 9 | 1370 | |||
GenBank mRNA | AF317220.1 | - | 4 | 307 | ||
AK001311.1 | - | 6 | 772 | |||
AK001428.1 | - | 6 | 826 | |||
AK310273.1 | - | 6 | 728 | |||
AY651263.1 | - | 6 | 755 | |||
BC019052.2 | - | 6 | 783 | |||
BC033349.1 | - | 6 | 999 | 4827210 | ||
U39317.1 | - | 6 | 395 |