Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300795.1 | NP_001287724.1 | 6, 7 | 486 | ||
NM_003340.6 | NP_003331.1 | 6, 7 | 793 | |||
NM_181886.3 | NP_871615.1 | 6, 7 | 1001 | |||
NM_181887.2 | NP_871616.1 | 6, 7 | 771 | |||
NM_181888.3 | NP_871617.1 | 6, 7 | 650 | |||
NM_181889.2 | NP_871618.1 | 6, 7 | 673 | |||
NM_181890.2 | NP_871619.1 | 6, 7 | 615 | |||
NM_181891.2 | NP_871620.1 | 6, 7 | 958 | |||
NM_181892.3 | NP_871621.1 | 5, 6 | 983 | |||
NM_181893.2 | NP_871622.1 | 5, 6 | 463 | |||
XM_017008584.1 | XP_016864073.1 | 5, 6 | 825 | |||
GenBank mRNA | AK095822.1 | - | 5, 6 | 898 | ||
AK127304.1 | - | 4, 5 | 3406 | |||
AK315775.1 | - | 6, 7 | 559 | |||
AL833414.1 | - | 5, 6 | 1576 | |||
AU131013.1 | - | 6, 7 | 561 | |||
BC003395.1 | - | 6, 7 | 473 | 3447673 | ||
BC037894.2 | - | 6, 7 | 770 | 5275356 | ||
BC066917.1 | - | 6, 7 | 504 | 4792462 | ||
BM541512.1 | - | 6, 7 | 755 | |||
BQ428985.1 | - | 6, 7 | 468 | |||
BU567649.1 | - | 6, 7 | 643 | |||
BU570661.1 | - | 5, 6 | 418 | |||
CR456916.1 | - | 5, 6 | 308 | |||
DA335600.1 | - | 6, 7 | 551 | |||
DA965437.1 | - | 5, 6 | 469 | |||
DB234695.1 | - | 5, 6 | 407 | |||
U39318.1 | - | 5, 6 | 353 |