Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300795.1 | NP_001287724.1 | 6 | 443 | ||
NM_003340.6 | NP_003331.1 | 6 | 750 | |||
NM_181886.3 | NP_871615.1 | 6 | 958 | |||
NM_181887.2 | NP_871616.1 | 6 | 728 | |||
NM_181888.3 | NP_871617.1 | 6 | 607 | |||
NM_181889.2 | NP_871618.1 | 6 | 630 | |||
NM_181890.2 | NP_871619.1 | 6 | 572 | |||
NM_181891.2 | NP_871620.1 | 6 | 915 | |||
NM_181892.3 | NP_871621.1 | 5 | 940 | |||
NM_181893.2 | NP_871622.1 | 5 | 420 | |||
XM_017008584.1 | XP_016864073.1 | 5 | 782 | |||
GenBank mRNA | AK095822.1 | - | 5 | 855 | ||
AK127304.1 | - | 4 | 3363 | |||
AK315775.1 | - | 6 | 516 | |||
AL833414.1 | - | 5 | 1533 | |||
AU131013.1 | - | 6 | 518 | |||
BC003395.1 | - | 6 | 430 | 3447673 | ||
BC037894.2 | - | 6 | 727 | 5275356 | ||
BC066917.1 | - | 6 | 461 | 4792462 | ||
BM541512.1 | - | 6 | 712 | |||
BQ428985.1 | - | 6 | 425 | |||
BU567649.1 | - | 6 | 600 | |||
BU570661.1 | - | 5 | 375 | |||
CR456916.1 | - | 5 | 265 | |||
DA335600.1 | - | 6 | 508 | |||
DA550074.1 | - | 6 | 362 | |||
DA965437.1 | - | 5 | 426 | |||
DB234695.1 | - | 5 | 364 | |||
U39318.1 | - | 5 | 310 |