Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103694868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773817.1 | XP_008772039.1 | 6 | 2391 |
Gene Symbol: LOC100910486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751788.3 | XP_003751836.2 | 6 | 2391 |
Gene Symbol: Slc27a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106145.1 | NP_001099615.1 | 10 | 1998 |