Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300795.1 | NP_001287724.1 | 7, 8 | 569 | ||
NM_003340.6 | NP_003331.1 | 7, 8 | 876 | |||
NM_181886.3 | NP_871615.1 | 7, 8 | 1084 | |||
NM_181887.2 | NP_871616.1 | 7, 8 | 854 | |||
NM_181888.3 | NP_871617.1 | 7, 8 | 733 | |||
NM_181889.2 | NP_871618.1 | 7, 8 | 756 | |||
NM_181890.2 | NP_871619.1 | 7, 8 | 698 | |||
NM_181891.2 | NP_871620.1 | 7, 8 | 1041 | |||
NM_181892.3 | NP_871621.1 | 6, 7 | 1066 | |||
NM_181893.2 | NP_871622.1 | 6, 7 | 546 | |||
XM_017008584.1 | XP_016864073.1 | 6, 7 | 908 | |||
GenBank mRNA | AK095822.1 | - | 6, 7 | 981 | ||
AK127304.1 | - | 5, 6 | 3489 | |||
AK315775.1 | - | 7, 8 | 640 | |||
AL833414.1 | - | 6, 7 | 1659 | |||
BC003395.1 | - | 7, 8 | 556 | 3447673 | ||
BC037894.2 | - | 7, 8 | 853 | 5275356 | ||
BC066917.1 | - | 7, 8 | 587 | 4792462 | ||
BU570661.1 | - | 6, 7 | 501 | |||
CR456916.1 | - | 6, 7 | 391 | |||
DA550074.1 | - | 7, 8 | 488 | |||
DA965437.1 | - | 6, 7 | 552 | |||
DB234695.1 | - | 6, 7 | 490 | |||
U39318.1 | - | 6, 7 | 436 |