Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106147.1 | NP_001099617.1 | 8 | 1321 | ||
XM_006254686.2 | XP_006254748.1 | 8 | 1549 | |||
XM_008772127.1 | XP_008770349.1 | 8 | 1549 |