Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152653.3 | NP_689866.1 | 2 | 315 | ||
XM_005265432.2 | XP_005265489.1 | 2 | 399 | |||
XM_005265433.4 | XP_005265490.1 | 2 | 399 | |||
XM_011534074.2 | XP_011532376.1 | 2 | 399 | |||
XM_011534075.1 | XP_011532377.1 | 2 | 399 | |||
XM_011534076.2 | XP_011532378.1 | 2 | 399 | |||
XM_017007126.1 | XP_016862615.1 | 2 | 399 | |||
GenBank mRNA | AK057886.1 | - | 2 | 316 | ||
AK307075.1 | - | 2 | 244 | |||
BC022332.1 | - | 2 | 291 | 4295679 |