Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202489.1 | NP_001189418.1 | 2 | 223 | ||
NM_003343.5 | NP_003334.2 | 4 | 305 | |||
NM_182688.2 | NP_872630.1 | 5 | 420 | |||
XM_017028469.1 | XP_016883958.1 | 4 | 264 | |||
GenBank mRNA | AF032456.1 | - | 4 | 272 | ||
AK290629.1 | - | 4 | 305 | |||
BC001738.1 | - | 4 | 267 | 3536853 | ||
BC008351.1 | - | 4 | 265 | 3536171 | ||
BC011569.1 | - | 4 | 277 | 4548763 | ||
BE292782.1 | - | 5 | 358 | |||
BT006914.1 | - | 4 | 217 | |||
DA710059.1 | - | 2 | 214 |