Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202489.1 | NP_001189418.1 | 4 | 456 | ||
NM_003343.5 | NP_003334.2 | 6 | 538 | |||
NM_182688.2 | NP_872630.1 | 7 | 653 | |||
XM_017028469.1 | XP_016883958.1 | 6 | 497 | |||
GenBank mRNA | AF032456.1 | - | 6 | 505 | ||
AK075143.1 | - | 2 | 679 | |||
AK122700.1 | - | 2 | 1955 | |||
AK290629.1 | - | 6 | 538 | |||
AL834129.1 | - | 2 | 1270 | |||
BC001738.1 | - | 6 | 500 | 3536853 | ||
BC008351.1 | - | 6 | 498 | 3536171 | ||
BC011569.1 | - | 6 | 510 | 4548763 | ||
BT006914.1 | - | 6 | 450 | |||
DA710059.1 | - | 4 | 447 |