Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100658.2 | NP_001094128.2 | 14 | 1765 | ||
GenBank mRNA | S75280.1 | - | 14 | 1712 |
Gene Symbol: LOC100912578 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749502.3 | XP_003749550.1 | 1 | 55 |