Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011910.1 | NP_001011910.1 | 8 | 924 | ||
GenBank mRNA | BC079381.1 | - | 8 | 924 | 7111053 | |
FQ218842.1 | - | 4 | 442 | |||
FQ235301.1 | - | 8 | 934 |