Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM189-UBE2V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199203.2 | NP_954673.1 | 8 | 1174 | ||
GenBank mRNA | BU156219.1 | - | 4 | 349 | ||
BU619725.1 | - | 2 | 147 | |||
CF145300.1 | - | 2 | 76 | |||
U39361.1 | - | 5 | 635 |