Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134840.1 | NP_001128312.1 | 20 | 2863 | ||
XM_008772271.1 | XP_008770493.1 | 20 | 3478 | |||
XM_008772272.1 | XP_008770494.1 | 20 | 3463 | |||
GenBank mRNA | BC168171.1 | - | 20 | 2805 |