Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134840.1 | NP_001128312.1 | 37 | 5496 | ||
XM_008772271.1 | XP_008770493.1 | 37 | 6111 | |||
XM_008772272.1 | XP_008770494.1 | 37 | 6096 | |||
GenBank mRNA | BC099150.1 | - | 6 | 765 |