Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003350.2 | NP_003341.1 | 3 | 289 | ||
XM_005251300.2 | XP_005251357.1 | 3 | 348 | |||
XM_011517583.2 | XP_011515885.1 | 3 | 367 | |||
XM_017013808.1 | XP_016869297.1 | 3 | 274 | |||
GenBank mRNA | AF049140.1 | - | 3 | 289 | ||
AK094617.1 | - | 2 | 1538 | |||
AK313447.1 | - | 3 | 292 | |||
BC007051.1 | - | 3 | 294 | 3683051 | ||
BC016332.1 | - | 3 | 314 | 4109228 | ||
BC016710.1 | - | 3 | 298 | 4108684 | ||
BC028673.1 | - | 3 | 291 | 4104992 | ||
BC062418.1 | - | 3 | 288 | 6503261 | ||
BT006744.1 | - | 3 | 268 | |||
CR407628.1 | - | 3 | 268 | |||
U62136.2 | - | 3 | 290 | |||
X98091.1 | - | 3 | 314 |