Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100945.1 | NP_001094415.1 | 2, 3 | 801 | ||
GenBank mRNA | AY724539.1 | - | 1, 2 | 444 | ||
BC129112.1 | - | 2, 3 | 801 | 7112017 |