Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC291863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013889.1 | NP_001013911.1 | 13 | 1357 | ||
GenBank mRNA | BC078681.1 | - | 13 | 1357 | 7123712 |
Gene Symbol: Ces1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133295.3 | NP_579829.3 | 12 | 1382 | ||
GenBank mRNA | AF171640.1 | - | 12 | 1331 | ||
BC061789.1 | - | 12 | 1382 | 5623732 | ||
CF111142.1 | - | 9 | 882 | |||
L46791.1 | - | 12 | 1361 | |||
X51974.1 | - | 12 | 1379 |