Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011925.1 | NP_001011925.1 | 14 | 1695 | ||
XM_008772282.1 | XP_008770504.1 | 12 | 3791 | |||
GenBank mRNA | BC081925.1 | - | 14 | 1695 | 7113037 |