Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnep1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106173.1 | NP_001099643.1 | 5 | 410 | ||
XM_008772374.1 | XP_008770596.1 | 4 | 345 |