Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005781.1 | NP_001005781.1 | 3, 4 | 317 | ||
NM_001005782.1 | NP_001005782.1 | 2, 3 | 242 | |||
NM_003352.4 | NP_003343.1 | 3, 4 | 317 | |||
GenBank mRNA | AB062294.1 | - | 3, 4 | 293 | ||
AK311840.1 | - | 3, 4 | 282 | |||
BC005899.1 | - | 3, 4 | 284 | |||
BC006462.1 | - | 3, 4 | 300 | 3546125 | ||
BC053528.1 | - | 3, 4 | 275 | 3452929 | ||
BC066306.1 | - | 3, 4 | 293 | 4818792 | ||
BG503431.1 | - | 3, 4 | 323 | |||
BG569821.1 | - | 2, 3 | 210 | |||
BT006632.1 | - | 1 | 169 | |||
BU532734.1 | - | 266 | ||||
CR542147.1 | - | 1 | 169 | |||
CR542156.1 | - | 1 | 169 | |||
U38784.1 | - | 4, 5 | 235 | |||
U61397.1 | - | 3, 4 | 314 | |||
U67122.1 | - | 3, 4 | 249 | |||
U72722.1 | - | 1 | 169 | |||
U83117.1 | - | 3, 4 | 304 | |||
X99586.1 | - | 210 |