Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam65a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100660.1 | NP_001094130.1 | 13 | 2258 | ||
XM_006255470.2 | XP_006255532.1 | 13 | 2607 | |||
XM_006255471.2 | XP_006255533.1 | 13 | 2719 | |||
XM_006255472.2 | XP_006255534.1 | 13 | 2708 | |||
XM_006255473.2 | XP_006255535.1 | 13 | 2608 | |||
XM_006255474.2 | XP_006255536.1 | 13 | 2607 | |||
XM_006255475.2 | XP_006255537.1 | 13 | 2271 | |||
GenBank mRNA | AY724537.1 | - | 5 | 1487 | ||
BC099098.1 | - | 3 | 1225 |