Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cirh1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009640.1 | NP_001009640.1 | 3 | 284 | ||
XM_006255562.1 | XP_006255624.1 | 3 | 330 | |||
XM_006255563.2 | XP_006255625.1 | 3 | 493 | |||
XM_008772488.1 | XP_008770710.1 | 3 | 283 | |||
GenBank mRNA | BC088461.1 | - | 3 | 284 | 7127580 |