Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106184.1 | NP_001099654.1 | 8, 9 | 1398 | ||
GenBank mRNA | BC168152.1 | - | 8, 9 | 1367 | 7461024 | |
JN712752.1 | - | 1410 |