Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024807.2 | NP_001019978.1 | 4 | 692 | ||
NM_005166.4 | NP_005157.1 | 4 | 692 | |||
XM_017026737.1 | XP_016882226.1 | 4 | 692 | |||
XM_017026738.1 | XP_016882227.1 | 4 | 692 | |||
GenBank mRNA | AK092376.1 | - | 4 | 492 | ||
AK297339.1 | - | 3 | 505 | |||
AK297640.1 | - | 4 | 391 | |||
AK312292.1 | - | 4 | 562 | |||
BC012889.1 | - | 4 | 569 | 3865417 | ||
BC013850.1 | - | 4 | 575 | |||
BC032983.1 | - | 4 | 565 | |||
U48437.1 | - | 4 | 541 |