Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx19a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005381.2 | NP_001005381.2 | 4 | 275 | ||
GenBank mRNA | AY303540.1 | - | 293 | |||
BC079094.1 | - | 4 | 246 | 7123989 | ||
CB582878.1 | - | 4 | 275 |
Gene Symbol: Ddx19b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005895.2 | NP_001005895.2 | 4 | 351 | ||
XM_008772534.1 | XP_008770756.1 | 4 | 265 | |||
GenBank mRNA | AY751756.1 | - | 4 | 293 | ||
EV774940.1 | - | 4 | 351 |